missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Mettler Toledo™ Accessories for FiveEasy™ Meters
For use with FiveEasy pH meters.
Manufacturer: Mettler Toledo™ 51302951
Description
- Can be attached to the left or right-hand side of the instrument
Specifications
pH meter | |
pH | |
Electrode Arm |