Mettler Toledo™ Filter Weighing Sets
For Micro and Ultramicro Series
Manufacturer:  Mettler Toledo™ 211227
 View more versions of this product
Catalog No. 01-913-77A
Includes: Dual plates to handle electrostatically charged films
Description
Description
  • In two styles to handle filters with diameters up to 50 and 110mm
  • Filter set for smaller diameter filters fits inside of (discontinued) UMT2 or MT5 weighing chamber, UMX, MX
  • Larger filter set mounts conveniently on top of weighing chamber of either balance
Specifications
Specifications
Filter Weighing Kit
For filters with maximum diameter of 110mm
Dual plates to handle electrostatically charged films
1 Set