Mettler Toledo™ Filter Weighing Sets
For Micro and Ultramicro Series
$1,768.00 - $3,660.80
Specifications
Includes Dual plates to handle electrostatically charged films
Quantity 1 Set
Product Type Filter Weighing Kit
View More Specs
Includes: Dual plates to handle electrostatically charged films
Products 2
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity  
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity  
01-913-77A
View Documents Promotion Details
Mettler Toledo™
211227
For filters with maximum diameter of 110mm
Each for $3,660.80
01-910-73
View Documents Promotion Details
Mettler Toledo™
211214
For filters with maximum diameter of 50mm
Each for $1,768.00
Description
Description
  • In two styles to handle filters with diameters up to 50 and 110mm
  • Filter set for smaller diameter filters fits inside of (discontinued) UMT2 or MT5 weighing chamber, UMX, MX
  • Larger filter set mounts conveniently on top of weighing chamber of either balance
Specifications
Specifications
Dual plates to handle electrostatically charged films
1 Set
Filter Weighing Kit