Please login to your online account to display your discounted pricing

Mettler Toledo™ FG2 FiveGo™ Portable pH Meter


Compact instrument for pH measurement in agriculture, food, beverage and water applications.

Manufacturer: mettler toledo™  ME51302900

 View more versions of this product

Catalog No. 01-915-655

  • / Each
 

Description & Specifications

Specifications

Display Type LCD
Weight (Metric) 0.18kg
Battery Type Four AAA
Temperature (Metric) Operating 0 to 40°C
Inputs BNC for pH combination, Cinch/RCA jack for ATC probe
Data Management Details 30 Measurements
Resolution (Temperature) 0.1°C
Range (pH) 0 to 14
Probe Type NTC
Temperature (Metric) Range -0° to +100°C
Temperature (English) Operating 32° to 104°F
Includes Meter, operating instructions, quick guide, electrode clip, wrist strap, four AAA batteries
Range (mV) 0 to ±1999mV
Resolution (pH) 0.01
Width (English) 3.2 in.
Material ABS/PC Reinforced Window, Membrane Keypad
For Use With (Application) pH
Height (Metric) 3.6cm
Length (Metric) 16.9cm
Weight (English) 0.4 lb.
Height (English) 1.4 in.
Calibration Points (pH) 1 or 2 Points, 3 pre defined buffer group
Resolution (mV) 1mV
Accuracy (pH) ±0.01
Accuracy (Temperature) ±0.5°C
Accuracy (mV) ±1mV