Please login to your online account to display your discounted pricing

Mettler Toledo™ FG2 FiveGo™ Portable pH Meter


Compact instrument for pH measurement in agriculture, food, beverage and water applications.

Manufacturer: mettler toledo™  ME51302902

 View more versions of this product

Catalog No. 01-915-657

  • / Each
 

Description & Specifications

Specifications

Includes Field Meter Kit: Meter, operating instructions, quick guide, electrode clip, wrist strap, four AAA batteries, LE438 pH/ATC electrode (01-915-675), IP54 Connection covers, 2 each pH 4.01 and pH 7 Buffer sachets, 4 Sample bottles, carry bag
Range (pH) 0 to 14
Resolution (pH) 0.01
Accuracy (pH) ±0.01
Range (mV) 0 to ±1999mV
Resolution (mV) 1mV
Accuracy (mV) ±1mV
Calibration Points (pH) 1 or 2 Points, 3 pre defined buffer group
Resolution (Temperature) 0.1°C
Accuracy (Temperature) ±0.5°C
Display Type LCD
Data Management Details 30 Measurements
Inputs BNC for pH combination, Cinch/RCA jack for ATC probe
Probe Type NTC
Battery Type Four AAA
IP Rating 54
Temperature (English) Operating 32° to 104°F
Temperature (Metric) Operating 0 to 40°C
Weight (English) 0.4 lb.
Weight (Metric) 0.18kg
For Use With (Application) pH
Height (English) 1.4 in.
Height (Metric) 3.6cm
Temperature (Metric) Range -0° to +100°C
Width (English) 3.2 in.
Material ABS/PC Reinforced Window, Membrane Keypad
Length (Metric) 16.9cm