Please login to your online account to display your discounted pricing

Mettler Toledo™ FE20 FiveEasy™ Benchtop pH Meter

Used for pH measurement in agriculture, food, beverage and water applications.  Mettler Toledo™ FE20 FiveEasy™ Benchtop pH Meter is a compact instrument that is versatile and easy to operate.

$837.72 - $1,044.43

Specifications

Accuracy (pH) ±0.01
Accuracy (Temperature) ±0.5°C
Calibration Points (pH) 1 or 2
Display Type LCD
Range (pH) 0 to 14
View More Specs
Includes:  Meter Only package comes with meter, operating instructions, Quick Guide, power adapter.
Meter Kit package includes meter, operating instructions, Quick Guide, power adapter, LE409 pH electrode (01-915-676), electrode arm, 2 each pH 4.01 and pH 7.00 buffer sachets
ATC Kit package includes meter, operating instructions, Quick Guide, power adapter, LE438 pH electrode (01-911-250), electrode arm, 2 each pH 4.01 and pH 7.00 buffer sachets
Products
Catalog Number Mfr. No. Includes Range (mV) Type Price Quantity    

01-915-661

 
mettler toledo™
ME51302910
Meter, operating instructions, quick guide 0 to 1999mV FE20 ph/mV/°C Meter Only Each for $837.72

01-915-662

 
mettler toledo™
ME51302911
Meter, operating instructions, quick Guide, power adapter, LE409 pH electrode (01-915-676), electrode arm, 2 each pH 4.01 and pH 7.00 buffer sachets 0 to 1999mV FE20 ph/mV/°C Meter Kit Each for $1,044.43
Description & Specifications

Specifications

Accuracy (pH) ±0.01
Accuracy (Temperature) ±0.5°C
Calibration Points (pH) 1 or 2
Display Type LCD
Range (pH) 0 to 14
Resolution (pH) 0.01
Height (English) 2.1 in.
Inputs BNC for pH combination or half cell, 2mm banana socket for separate reference half cell, Cinch/RCA jack for ATC probe (NTC 30kilohm)
Length (Metric) 20cm
Material ABS/PC Reinforced Window, Membrane Keypad
Outputs 2mm Banana socket for analog output
Probe Type NTC
Resolution (mV) 1mV
Resolution (Temperature) 0.1°C
Temperature Range (English) -32° to +212°F
Temperature Range (Metric) -0° to +100°C
Voltage 100/240V
Weight (English) 1.3 lb.
Weight (Metric) 0.6kg
Width (Metric) 17.5cm

Powerful, time-saving features

  • Automatic or manual termination of a measurement or calibration
  • Automatic calibration with predefined pH buffer groups
  • Automatic or manual temperature compensation
  • Two-point calibration with automatic buffer recognition
Large and informative LCD
  • Simultaneous display of measurement, temperature and endpoint
  • Electrode icon provides information on electrode condition
  • Calibration/measurement icon shows when instrument is calibrating or carrying out measurements
  • Setup icon lets you enter and modify settings
Easy operation
  • Simple start and finish of measurements and calibrations
  • Fast changeover between pH and mV parameters
  • Intuitive adjustment of settings such as manual temperature compensation
  • Save and access measurements and calibration data at the push of a button
  • Separate on/off switch
  • Measures pH, mV and temperature
  • Laminated Quick Guide outlines operation at a glance

Variety of inputs and outputs

  • BNC input for combination or half-cell electrode
  • Reference input for separate reference half cell
  • Accepts redox electrode for oxidation-reduction measurements
  • ATC input for NTC 30kω probe
Durable housing, universal power supply
  • Rugged ABS housing
  • PC-reinforced window
  • Tough membrane keypad
  • 100 to 240V 50/60Hz power supply provides 9VDC to instrument