missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mettler Toledo™ F1 Premium Weight Sets with Certificate

Made of vacuum melted steel for highest material purity

$2,371.50 - $2,371.50

Specifications

Certifications/Compliance With certificate, ISO17025, ISO9001, ISO14001 environmental standard
Classification OIML Class F1
Density 8kg/dm3 at 20°C
Material High Grade Stainless Steel
Product Type Weight set
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Includes Quantity Weight (Metric) Price Quantity  
Catalog Number Mfr. No. Includes Quantity Weight (Metric) Price Quantity  
01-911-669 Mettler Toledo™
ME158907
Wire weight: 1mg, 2mg, 5mg, 10mg, 20mg, 50mg, 100mg, 200mg, 500mg, marked wire weight: 2mg, 20mg, 200mg, cylindrical weight: 1g, 2g, 5g, 10g, 20g, 50g, 100g, 200g, marked cylindrical weight: 2g, 20g, 200g, Plastic box 13/Set 1mg to 200g Each for $2,371.50
Specifications

Description

Thanks to their monobloc design, Mettler Toledo Premium Line weights are used in mass labs all over the world, and for calibration of analytical and micro balances. Premium, vacuum melted steel offers an anticorrosive surface, and low magnetization and susceptibility values. Monobloc design and electro polishing of surface provide maximum stability. Initial weight calibration uses robotic technology which excludes human error and leads to consistent and reproducible results with low uncertainty values.

  • Premium Line OIML F1 weight sets
  • 1mg - 500mg: wire weights
  • 1g - 5kg: cylindrical knob weights
  • High-grade steel
  • Density: 8.0 kg/dm3
  • Magnetic susceptibility: <0.01
  • Impact-resistant plastic box
  • Knob weights: Monobloc
  • Second weight of same mass is marked for identification purposes

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.

Description & Specifications

Specifications

With certificate, ISO17025, ISO9001, ISO14001 environmental standard
8kg/dm3 at 20°C
Weight set
OIML Class F1
High Grade Stainless Steel
Multiple