Please login to your online account to display your discounted pricing

Mettler Toledo™ F1 Premium Cylindrical Knob Weights with Certificate


Individual weights

Manufacturer: mettler toledo™  ME158667

 View more versions of this product

Catalog No. 01-911-659

  • / Each
 

Description & Specifications

Specifications

Includes Plastic box
Classification OIML Class F1
Packaging Quantity 1
Density 8kg/dm3 at 20°C
Material High Grade Stainless Steel
Weight (Metric) 100g
Mass 100g
Shape Cylindrical
Product Type Weight with knob, One piece (monobloc)
Quantity Each
Certifications/Compliance With certificate, ISO17025, ISO9001, ISO14001 environmental standard