Please login to your online account to display your discounted pricing
Mettler Toledo™ Excellence XSE Analytical Balances
Worry-free weighing thanks to intuitive operation
$8,543.25 - $8,543.25
Specifications
Electrical Requirements 110/230V 50/60Hz
Certifications/Compliance GLP, ISO 9001, UL, CSA
View More Specs
Products
Catalog Number Mfr. No. Capacity (Metric) Readability Repeatability Price Quantity    
01-911-007
mettler toledo™
ME30087708
120g 0.1mg <±0.4mg Each for $8,543.25
01-911-006
mettler toledo™
ME30087732
81 / 220g 0.01 / 0.1mg <±0.1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
01-911-005
  Accessories
mettler toledo™
ME30087716
41 / 120g 0.01 / 0.1mg <±0.1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
01-911-008
  Accessories
mettler toledo™
ME30087724
220g 0.1mg <±0.4mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications
Specifications
Electrical Requirements 110/230V 50/60Hz
Certifications/Compliance GLP, ISO 9001, UL, CSA
XSE Analytical Balances offer exceptional weighing performance and quality assurance features as well as excellent ergonomics. They are the perfect solution for a multitude of workplaces. Graphical LevelControl and MinWeigh™ enable full regulatory compliance. Innovative StatusLight™ feature takes the worry out of weighing. A green light lets you know when it's safe to start, indicating all tests are up-to-date and balance is level and functioning correctly.
  • StatusLight uses color to help you identify the status of your balance, so you know exactly when it is ready for use
  • SmartGrid™ weighing pan minimizes effects of turbulence in weighing chamber; ensures more accurate results and shorter stabilization times
  • SmartScreen™ display offers color-guided touchscreen for safe operation
  • Individual, color-coded profiles can be configured and documented for up to eight users or jobs, protecting users, data and processes, and guaranteeing seamless traceability
  • ErgoClips™ facilitate secure placement and efficient handling of tare containers
  • LevelControl warns if balance is not correctly leveled
  • MinWeigh signals when sample has not reached minimum weight defined by regulations
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.