Mettler Toledo™ Excellence, XS Series Precision Balances
Designed for uncompromising efficiency in laboratory and industrial environments. Mettler Toledo Excellence, XS Series Precision Balances deliver all the information needed for Good Laboratory Practice. All balances come with full range taring.
$5,320.46 - $8,919.06
Specifications
Material Die-cast Aluminum, Plastic, Chrome Steel
Electrical Requirements 100-240V AC, 50/60 Hz, 12V DC ±3%, 2.5 A
Certifications/Compliance UL Listed. CE compliant.
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Model Stabilization Time Capacity (Metric) Readability Price Quantity  
Catalog Number Mfr. No. Model Stabilization Time Capacity (Metric) Readability Price Quantity  
01-913-887 Mettler Toledo™
ME30134079
XS303S 1.5 sec. 310g 1mg Each for $6,324.49
01-913-888 Mettler Toledo™
ME30134167
XS1202S 1.2 sec. 1210g 10mg Each for $5,320.46
01-913-889 Mettler Toledo™
ME30134179
XS1203S 1.5 sec. 1210g 1mg Each for $8,280.76
01-913-890 Mettler Toledo™
ME30134197
XS10002S 1.5 sec. 10.1kg 10mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
01-913-891 Mettler Toledo™
ME30134205
XS10001L 1 sec. 10.1kg 100mg Each for $8,454.10
01-914-058 Mettler Toledo™
ME30265590
XS303SN 1.5 sec. 310g 1mg Each for $6,098.55
01-914-059 Mettler Toledo™
ME30265602
XS603SN 1.5 sec. 610g 1mg Each for $8,919.06
Specifications
Description
Operating menu can be viewed in English, German, French, Spanish, Italian, Russian, and Japanese. All models can download software from the Internet, display various units of weight, tare over entire weighing range, display statistics with ± and %, count with reference optimization, perform formula weighing, and calculate with factors.
The innovative SmartPan™ helps you to get your work done more quickly by delivering accurate and stable results two times faster, even in difficult weighing environments such as fume cupboards and safety cabinets. Included as standard on all 0.01 g balances, the SmartPan weighing pan minimizes the effects of air currents on the weighing cell. All units have backlit graphic display with touch screen operation, fully automatic internal adjustment, alphanumeric sample ID entry, and IP54 in-use protection. The following optional interfaces are available: Bluetooth™ (wireless), Ethernet, PS/2, USB, LocalCAN™, or MiniMettler. Other peripherals are available; contact your sales representative for more information. Balance applications include statistics, percent weighing, animal/dynamic weighing, piece counting, formulation, and density determination. Electrical: 115/230V, 50/60Hz.
  • Capacities from 210g to 32.1kg
  • Readabilities from 0.001 to 1g
  • Features proFACT™ Fully Automatic Calibration Technology
  • Meets All GLP and ISO 9001 Requirements
Compliance
METTLER TOLEDO provides ISO 9001 manufacturing and nationwide ISO 9001 field service. UL listed and CSA certified.
Description & Specifications
Specifications
Die-cast Aluminum, Plastic, Chrome Steel
100-240V AC, 50/60 Hz, 12V DC ±3%, 2.5 A
UL Listed. CE compliant.