Mettler Toledo™ Excellence, XS Series Analytical Balances
Offers a variety of uses in pharmaceutical, chemical, food, cosmetic, research and development, and quality control applications
$9,528.82 - $9,528.82
Specifications
Electrical Requirements 100-240V AC, 50/60 Hz, 12V DC ±3%, 2.5 A
Certifications/Compliance UL Listed. CE compliant.
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Model Stabilization Time Capacity (Metric) Readability Price Quantity  
Catalog Number Mfr. No. Model Stabilization Time Capacity (Metric) Readability Price Quantity  
01-913-892 Mettler Toledo™
ME30132870
XS105 3 sec. 120g 0.01mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
01-913-893 Mettler Toledo™
ME30132858
XS225DU 1.5 / 3.0 sec. 120 / 220g 0.1 / 0.01mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
01-913-894 Mettler Toledo™
ME30132948
XS304 1.5 sec. 320g 0.1mg Each for $9,528.82
Specifications
Description
XS Series analytical balances feature the unique SmartGrid™ weighing pan, which is supported at the rear of the weighing chamber, for more efficient weighing. The SmartGrid weighing pan has only minimal solid area to minimize buoyancy caused by turbulance, shortening stabilization time for faster results. In combination with the optional weighing kit, which includes ErgoClips™, the SmartGrid weighing pan becomes an ergonomic weighing system that can be precisely adapted to specific tasks and can help to increase productivity. ErgoClips™ click securely onto the grid weighing pan and can hold round-bottom flasks, test tubes, plastic tubes, and more.
All units have backlit graphic display with touch-screen operation, fully automatic internal adjustment, and a built-in RS-232 interface. These balances make cleaning easy with a draft shield that can be completely dismantled. The dishwasher-safe inner base plate and SmartGrid weighing pan can also be removed for easy cleaning. Peripherals can be connected via the built-in RS-232 interface or an optional second interface (sold separately). The following optional interfaces are available: Bluetooth™ (wireless), Ethernet, PS/2, USB, LocalCAN™, or MiniMettler.
  • Capacities from 61 to 220g
  • Readabilities from 0.1 to 0.01mg
  • Features FACT™ Fully Automatic Calibration Technology
  • Meets All GLP and ISO 9001 Requirements
  • Built-in RS-232 Interface
  • Bluetooth™ Wireless Technology available
Compliance
METTLER TOLEDO provides ISO 9001 manufacturing and nationwide ISO 9001 and ISO 17025/A2LA certified field service. UL listed and CSA certified.
Description & Specifications
Specifications
100-240V AC, 50/60 Hz, 12V DC ±3%, 2.5 A
UL Listed. CE compliant.