Please login to your online account to display your discounted pricing
Mettler Toledo™ ErgoClips™ Solution Kits
Allow easy weighing directly into tare containers. Mettler Toledo ErgoClips Solution Kit includes all necessary tools for direct dosing with storage case.
Manufacturer: mettler toledo™  ME11140253
 View more versions of this product
Catalog No. 09-320-360
  • / Each
Description & Specifications
Specifications
Product Type ErgoClip™ Solution Kit
Includes Min weigh micro door, tube, micro flask, including extension, micro basket, micro stand, SmartGrid™, tweezers, 5mL volumetric flasks, small reagent tube, 1.5 and 0.5mL centrifuge tube, 2mL HPLC tube, micro spatula
For Use With (Equipment) XP26 and XP56 Micro balance
Type ErgoClip™ solution kit
  • Guarantees secure placement of different tare containers
  • Dosing is fast and safe thanks to the adjustable MinWeight Door
  • Accurate weighing for reliable, complete sample transfer
Solution Kit Contents:
  • ErgoClip Flask
  • ErgoClip Tube
  • ErgoClip Flask (Small)
  • ErgoClip Round Bottom Flask
  • ErgoClip Basket (Small)
  • ErgoClip Weighing Boat
  • ErgoClip Titration Basket
  • ErgoClip Quantos and vial adapters
  • ErgoClip Filter holder
  • MinWeight Door
  • Stand
  • Various tare containers
  • Spatula
  • Tweezers
Solution Kit Micro Contents:
  • MinWeight Door micro
  • ErgoClip Tube (without holder)
  • ErgoClip Flask micro, including extension
  • ErgoClip Basket micro
  • Stand for ErgoClips micro
  • SmartGrid
  • Tweezers
  • 5mL volumetric flasks
  • Reagent tube, small
  • 1.5 and 0.5mL centrifuge tube
  • 2mL HPLC tube
  • Micro spatula
Solution kit for use with analytical balances.
Micro kit for use with micro balances XP26 or XP56.