Mettler Toledo™ EL2 Portable pH Meter for Teaching and Learning
Compact instrument tailored for field experiments and training courses
in colleges and universities.
Manufacturer:  Mettler Toledo™ ME51302922
 View more versions of this product
Catalog No. 01-915-667
Specifications Description & Specifications
Specifications
Field Kit: Meter, operating instructions, quick guide, electrode clip, wrist strap, four AAA batteries, LE407 pH/ATC electrode (01-915-675), IP54 Connection covers, 2 each pH 4.01 and pH 7 buffer sachets, 4 sample bottles, carry bag
0.01
0 to ±1999mV
±1mV
0.1°C
LCD
NTC
54
0 to 40°C
1.4 in.
-0° to +100°C
16.9cm
0 to 14
±0.01
1mV
1 or 2
±0.5°C
Current calibration, 30 measurements
Four AAA
32° to 104°F
pH
3.6cm
3.2 in.