Mettler Toledo™ Accessories for EasyPlus™ Titrator
For use with Mettler Toledo EasyPlus Titrators.
Manufacturer: Mettler Toledo™ 30065504
Description
- Vpac Chloride
- Sodium chloride (NaCl)
- Titrant: AgNO3 0.1 mol/L
- For performance verification
Specifications
Easy Cl and Easy Pro Titrators | |
Chloride Titration Standard (Sodium chloride (NaCl) |