Mettler Toledo™ EasyPlus™ Chloride, Redox mV, and Redox Ipol and Karl Fischer™ Sensors
Specially developed for use with the EasyPlus Titrators
Manufacturer: Mettler Toledo™ 30043105
Description
- Used for chloride precipitation, redox titrations/voltammetric/amperometric titrations
- Delivers quality and reliable results
- Made of glass
Chloride Sensor
- Silver ring sensor for chloride titrations (especially for food and water)
- Also used in silver content titrations
- Platinum ring sensor for redox titrations (especially for food, electoplating or electronic industries)
- Also used in applications like peroxide value or reducing sugar content
- Simple, durable and zero-maintenance sensor primarily for Karl Fischer and redox titrations
- For applications like important food ingredients such as Vitamin C or SO2 in wine
Specifications
BNC Fixed Cable | |
Glass | |
0°C to 100°C |
Karl Fischer Redox titration | |
0°F to 212°F | |
Titration Sensor |