Please login to your online account to display your discounted pricing

Mettler Toledo™ E2 Premium Weight Sets with Certificate


Made of vacuum melted steel for highest material purity

Manufacturer: mettler toledo™  ME11117322

 View more versions of this product

Catalog No. 01-911-854

  • / Each
 

Description & Specifications

Specifications

Product Type Weight set
Certifications/Compliance With certificate, ISO17025, ISO9001, ISO14001 environmental standard
Includes Wire weight: 1mg, 2mg, 5mg, 10mg, 20mg, 50mg, 100mg, 200mg, 500mg, marked wire weight: 2mg, 20mg, 200mg, cylindrical weight: 1g, 2g, 5g, 10g, 20g, 50g, 100g, 200g, 500g, 1kg, 2kg, marked cylindrical weight: 2g, 20g, 200g, 2kg, Plastic box
Classification OIML Class E2
Quantity 27/Set
Packaging Quantity 27
Material High Grade Stainless Steel
Weight (Metric) 1mg to 2kg
Density 8kg/dm3 at 20°C
Mass 1g to 2kg