missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Mettler Toledo™ DL31 Karl Fischer™ Titrator Replacement Parts: Electrodes
Replacement parts for DL31 Karl Fischer titrators.
Manufacturer: Mettler Toledo™ 51107699
Description
- For use with DL31 Karl Fischer titrators
- Type: Double-Pin Platinum Electrode
- For voltametric titrations
Specifications
For voltametric titration | |
144.5 mm | |
0°F to 176°F | |
Double-Pin Platinum Electrode |
5.6 in. | |
Platinum | |
0°C to 80°C |