Mettler Toledo™ Detachable Cables for S7 Screw Head Electrodes
Connects electrodes with an S7 screw head to meters with specified connector. Mettler Toledo Detachable Cables can be used with meters that have a BNC, 2mm Banana or US Standard connector.
Manufacturer: Mettler Toledo™ 52300016
Description
- Detachable cables that attach to electrodes with an S7 screw head
- Length 1.2m (3.9 ft.)
Specifications
2mm Banana | |
1.2m | |
Cable |
47.2 in. | |
S7 Screw head electrode with 2mm banana connector |