Please login to your online account to display your discounted pricing
Mettler Toledo™ Density Kit
For use with XS/XP balances. Mettler Toledo Density Kits helps to determine the density of solids and liquids with top-loading electronic balances.
Manufacturer: mettler toledo™  ME11132680
Catalog No. 09-320-104
  • / Each
Description & Specifications
Specifications
For Use With (Equipment) XS/XP balance
Includes Solids kit includes 200mL beaker, thermometer, bridge, sample holder (2), bracket, 25mL bottle of wetting agent, liquids kit includes sinker mounted with pan and Platinum wire
Solids Kit
  • 200mL beaker
  • Thermometer
  • Bridge
  • Sample Holder (2)
  • Bracket
  • Bottle of wetting agent, 25mL
Liquids Kit
  • Sinker mounted with pan and platinum wire
Note
Both kits are required to perform liquid density measurements.
Made to order. Non-returnable.