Please login to your online account to display your discounted pricing
Mettler Toledo™ Computer Data Transfer Cables
For AT, BD Series; MT5 and UMT2 Balances.
Manufacturer: mettler toledo™  ME210491
 View more versions of this product
Catalog No. 01-910-15
  • / Each
Description & Specifications
Specifications
For Use With (Equipment) IBM XT/PS2, AX and MX UMX balance
Length (English) 59.05cm
Length (Metric) 23.24 in.
Connection Type 15 Pin, 25 socket, RS232
  • All link AT, UMT2, and MT5 Series balances to selected computers via the standard RS-232-C/CL interface (09-317-3 also works with BD Series balances)
  • Allow data collection by computer