Please login to your online account to display your discounted pricing
Mettler Toledo™ Classic Light Series Balances
Ideal for simple weighing tasks; compact, economical
$3,721.31 - $3,721.31
Products
Catalog Number Mfr. No. Price Quantity    
01-910-154
mettler toledo™
ME12120512
Each for $3,721.31
Description & Specifications
  • Simple operation with logical menu guidance and only few keys
  • Automatic adjustment with external weight and automatic shutoff
  • Large, bright, seven-segment LCD readout Built-in overload protection
  • Large weighing pan for maximum working efficiency
  • Built-in below-balance weighing capability
  • AC adapter appropriate to national standard
AL-Models
  • Units include g, kg, mg, ct, lb., oz., ozt, GN, dwt, momme, mesghal, tael, tical, tola, and baht
  • With glass draft shield that opens to sides and top
PL Models
  • Weighing modes: g, kg, mg, carats, lb., oz., ounces troy, grains, pennyweight, mommes, mesghals, taels (three variants), ticals, tolas and bahts
  • PL-S models offer the option of a built-in rechargeable storage battery
Fisher Service Division offers several types of service agreements for the equipment featured here. For details on these agreements, equipment repairs, and calibration services, see the front of the catalog. For immediate assistance, call 1-800-395-5442.
AL- models: Electrical Requirements: 120V 50/60Hz. Use with line cord (included) or four “AA” batteries.
PL-models: Electrical Requirements: 105/120V 50/60Hz. A variety of accessories are available on www.fishersci.com including optional carrying case and in-use covers.
Service agreement available.
One year