Please login to your online account to display your discounted pricing
Mettler Toledo™ Classic Light Series Balances
Ideal for simple weighing tasks; compact, economical
Manufacturer: mettler toledo™  ME12120512
 View more versions of this product
Catalog No. 01-910-154
  • / Each
Description & Specifications
Specifications
Capacity (Metric) 210g
Readability 0.0001g
Type Analytical Balance
  • Simple operation with logical menu guidance and only few keys
  • Automatic adjustment with external weight and automatic shutoff
  • Large, bright, seven-segment LCD readout Built-in overload protection
  • Large weighing pan for maximum working efficiency
  • Built-in below-balance weighing capability
  • AC adapter appropriate to national standard
AL-Models
  • Units include g, kg, mg, ct, lb., oz., ozt, GN, dwt, momme, mesghal, tael, tical, tola, and baht
  • With glass draft shield that opens to sides and top
PL Models
  • Weighing modes: g, kg, mg, carats, lb., oz., ounces troy, grains, pennyweight, mommes, mesghals, taels (three variants), ticals, tolas and bahts
  • PL-S models offer the option of a built-in rechargeable storage battery
Fisher Service Division offers several types of service agreements for the equipment featured here. For details on these agreements, equipment repairs, and calibration services, see the front of the catalog. For immediate assistance, call 1-800-395-5442.