Please login to your online account to display your discounted pricing
Mettler Toledo™ C20 and V20 Series Karl Fischer™ Titrators
Simplest, most secure water determination. Mettler Toledo C20 and V20 Series Karl Fischer Titrators provide water determination running with one click of a touchscreen.
Specifications
mV Range -2000 to 2000mV
Resolution 0.1mV
Temperature Range (English) Ambient 41° to 104°F
Temperature Range (Metric) Ambient 5° to 40°C
View More Specs
Products
Catalog Number Mfr. No. Detection Range Model Type Price Quantity    
01-911-136
mettler toledo™
ME51105512
1ppm to 5% C20X (without Diaphragm) Coulometric titrator CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
01-911-137
mettler toledo™
ME51105510
1ppm to 5% C20D (with Diaphragm) Coulometric titrator CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications
Specifications
mV Range -2000 to 2000mV
Resolution 0.1mV
Temperature Range (English) Ambient 41° to 104°F
Temperature Range (Metric) Ambient 5° to 40°C
One Click solvent exchanges ease the task of emptying and filling the vessel—and all other daily tasks can be simplified to One Click Shortcuts on the Home Screen. True Plug-and-Play technology adds automatic recognition of burets on the V20, further enhancing security. 
  • Simple and fast operation
  • Safe handling of chemicals
  • Easy-to-use touchscreen interface
  • User-specific Home Screen featuring intuitive shortcuts for routine tasks
  • Coulometric generation cell; choose Model C20D with diaphragm or Model C20X without diaphragm
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.