Please login to your online account to display your discounted pricing

Alfa Aesar™ Metronidazole, 50mg/mL in 0.1N HCl, sterile-filtered


Quantity 25mL

Manufacturer: alfa aesar™  J67362AC

 View more versions of this product

Catalog No. AAJ67362AC

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

CAS 443-48-1
Chemical Name or Material Metronidazole
Physical Form Liquid
MDL Number MFCD00009750
Health Hazard 1 Warning
Storage Note 1 -30° to -10°C
Quantity 25mL
Name Note 50mg/mL in 0.1N HCl, sterile-filtered
Health Hazard 2 GHS H Statement
H351
Suspected of causing cancer.
RTECS Number NI5600000
Health Hazard 3 GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Specifications

Chemical Name or Material Metronidazole
CAS 443-48-1
Name Note 50mg/mL in 0.1N HCl, sterile-filtered
MDL Number MFCD00009750
Accessories

Warning

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H351
Suspected of causing cancer.

-30° to -10°C