Metrohm Titrode™ Redox Metal Combination Electrodes
Unique electrodes combine glass and noble metal electrodes into a single assembly.
$1,493.18 - $1,493.18
Products ${productFamilyLength}
Catalog Number Mfr. No. Price Quantity  
Catalog Number Mfr. No. Price Quantity  
BR20924004 Metrohm
020924004
Each for $1,493.18
Specifications
Description
No reference diaphragm; therefore, no filling solution or diaphragm-related problems. Electrodes are available in platinum (for redox reactions), silver (for precipitation [argentometry] reactions), and double platinum sheet (for low-level redox reactions).
  • Temperature range, 0° to 80°C (32° to 176°F); -20° to +70°C (-4° to +158°F) for double platinum sheet
  • Easily stored in distilled water
For pH titrations only, not for direct measurements