Metro™ Wire Shelving Track Sets
For use with Metro Super Erecta™, Super Erecta Pro™ and Super Adjustable™ Top Track™ shelving
Manufacturer:  Metro™ TTS16NA
 View more versions of this product
Catalog No. 19-138-239
Description
Description
  • Track sets include necessary sections of track for assembling track runs up to 21 ft. (640cm)
  • Coated with Microban™ antimicrobial protection
Order Info
19-138-228 (TTS2.5NA) includes a joining kit to connect the track to other track sets.
Specifications
Specifications
Metro™ Super Erecta™, Super Erecta Pro™, Super Adjustable™ top track shelving
Necessary sections of track for assembling track runs of 1830 to 6405mm
192 in.
487.7cm
Documents