Metro™ Type 304 Stainless Steel Garment Racks
Open wire construction promotes laminar air flow, Metro Type 304 Stainless Steel Garment Racks have unique hanger capture mechanism that provides theft deterrance.
Manufacturer: Metro™ GRW2448S
Description
- Type 304 Stainless steel and Proven Super Erecta™ construction throughout
- Floor-mountable upright and wall mount versions available
- Open wire construction promotes laminar air flow
- Electro-polished, Type 304 Stainless steel hangers are included. Hangers are spaced at 7.62cm (3 in.) intervals
- Unique hanger capture mechanism provides theft deterrance
- Easy to assemble
- Adjustable stainless steel triangular foot plates provided for floor mounting
- Upright unit is non-conductive, but can be made conductive with the addition of aluminum split sleeves (9986Z)
- Floor Mount style is 189.5cm (74.625 in.) tall. Wall Mount style is 19.05cm (7.5 in.) high
Specifications
48 x 24 in. (122 x 61cm) | |
122cm | |
61cm |
48 in. | |
24 in. |