Metro™ Super Erecta™ Wire Shelf, Stainless Steel Finish
Transform your laboratory or workplace storage as you see fit. Metro Super Erecta Wire Shelves in stainless steel finish allow adjustable configurations when used with Metro Super Erecta shelving systems
Supplier:  Metro™ 2436NS
 View more versions of this product
Catalog No. 19-138-742
Description
Description
  • Open-wire design minimizes dust accumulation, allows free circulatin of air and greater visibility of stored items
  • Type 304 polished stainless steel
  • Used for the most aggressive applications and environments
  • Stands up to high temperature automated cart wash and autoclave applications
Specifications
Specifications
61 x 91.4 in. (24 x 36 cm)
24 in.
24 in.
61 cm
91.4 cm
Plastic sleeves and wedges
36 in.
610 mm
Metro™ Super Erecta™ shelving system
Stainless Steel