Metro™ MetroMax i™ Lab Worktable, Stainless Top and Backsplash with Solid i Shelf
The Metro MetroMax i™ Lab Worktable advanced polymer construction provides excellent corrosion-resistance performance for the most corrosive lab environments.
Manufacturer:  Metro™ LTM48XS3
 View more versions of this product
Catalog No. 11-100-238
Specifications Description & Specifications
Specifications
Stainless Steel
Ideal for lab
100.3cm
47.75 in.
600 lb.
Polymer
30 in.
47.75 x 30 x 39.5 in. (121.3 x 76.2 x 100.3cm)
39.5 in.
Backsplash with solid Metro™ MetroMax i™ shelf, adaptor insert
121.3cm
273kg
Mobile Table
76.2cm