Metro™ MetroMax i™ Lab Worktable, Stainless Top and Backsplash with 3-Sided Frame
The Metro MetroMax i™ Lab Worktable advanced polymer construction provides excellent corrosion-resistance performance for the most corrosive lab environments.
Manufacturer:  Metro™ LT36XUS3
 View more versions of this product
Catalog No. 11-100-211
Specifications Description & Specifications
Specifications
Stainless Steel
Ideal for lab
100.3cm
35.75 in.
600 lb.
Polymer
30 in.
35.75 x 30 x 39.5 in. (90.8 x 76.2 x 100.3cm)
39.5 in.
Backsplash with 3 sided frame
90.8cm
273kg
Stationary Table
76.2cm