Please login to your online account to display your discounted pricing

Metro™ MetroMax i Glassware Cart, Accessories


For use with MetroMax i carts

Manufacturer: metro™  GWOLYKIT

 View more versions of this product

Catalog No. 13-451-099

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) For easy and efficient way to transport laboratory Glassware
Color Gray
For Use With (Equipment) For MetroMax Glassware carts
Includes 6 in. Deep Metrotote box, two long divider, two short divider
Material Polypropylene
Product Type Glassware transport kit