Please login to your online account to display your discounted pricing

Metolazone, Fisher BioReagents


Quantity 50mg
Packaging Amber Glass

Manufacturer: fisher scientific company  BP266750

 View more versions of this product

Catalog No. BP266750

  • / Each
  • / Case of 15 Each
 

Description & Specifications

Chemical Identifiers

FTIR Conforms to reference
Packaging Amber Glass
Color White
Melting Point 248°C / °F
Quantity 50mg
Chemical Name or Material Metolazone
CAS 17560-51-9
Physical Form Powder; Solid
Molecular Formula C16H16ClN3O3S
Formula Weight 365.8g/mol
Merck Index 15, 6224
Synonym Zaroxolyn
ChemAlert Storage Symbol Gray
Recommended Storage RT

Specifications

Chemical Name or Material Metolazone
CAS 17560-51-9
Physical Form Powder; Solid
Molecular Formula C16H16ClN3O3S
Formula Weight 365.8g/mol
Synonym Zaroxolyn

Metolazone is an anti-hypertensive.

Accessories
 

RT

SDS & Safety Documents