Please login to your online account to display your discounted pricing

Metolazone, Fisher BioReagents

$90.49 - $90.49

Chemical Identifiers

Chemical Name or Material Metolazone
CAS 17560-51-9
Physical Form Powder/Solid
Molecular Formula C16H16ClN3O3S
Formula Weight 365.8
Synonym Zaroxolyn
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

BP266750

  SDS
fisher scientific company
BP266750
Amber Glass 50mg Each for $90.49
Case of 15 Each for $1,357.35
Description & Specifications

Specifications

Physical Form Powder/Solid
Formula Weight 365.8
Melting Point 248°C
Quantity 50mg
Merck Index 15, 6224
Molecular Formula C16H16ClN3O3S
Color White
Chemical Name or Material Metolazone
Packaging Amber Glass
CAS 17560-51-9
ChemAlert Storage Symbol Gray
Synonym Zaroxolyn
Recommended Storage RT
FTIR Conforms to reference

Metolazone is an anti-hypertensive.

Safety and Handling
 

RT