missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methylurea, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2477230
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Methylurea | |
96°C to 101°C | |
Odorless | |
MFCD00007950 | |
878189 | |
XGEGHDBEHXKFPX-UHFFFAOYSA-N | |
methylurea | |
11719 | |
74.08 |
598-50-5 | |
6.7 | |
C2H6N2O | |
250 g | |
1-methylurea, n-methylurea, monomethylurea, urea, methyl, methyl urea, urea, n-methyl, methylmocovina, methylmocovina czech, methylharnstoff german, n-methyl urea | |
CNC(=O)N | |
74.083 | |
CHEBI:44383 | |
98% |
Chemical Identifiers
598-50-5 | |
74.083 | |
XGEGHDBEHXKFPX-UHFFFAOYSA-N | |
11719 | |
methylurea |
C2H6N2O | |
MFCD00007950 | |
1-methylurea, n-methylurea, monomethylurea, urea, methyl, methyl urea, urea, n-methyl, methylmocovina, methylmocovina czech, methylharnstoff german, n-methyl urea | |
CHEBI:44383 | |
CNC(=O)N |
Safety and Handling
EINECSNumber : 209-935-0
RTECSNumber : YT7175000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only