missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyltriphenylphosphonium iodide, 98%

$68.50 - $68.50

Chemical Identifiers

CAS 2065-66-9
Molecular Formula CH3P(C6H5)3I
Formula Weight 404.23
MDL Number MFCD00066175
UN Number UN3464
Chemical Name or Material Methyltriphenylphosphonium iodide
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1564414 SDS Alfa Aesar™
A1564414*
25g Each for $68.50
AAA1564422 SDS Alfa Aesar™
A1564422
100g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAA1564436 SDS Alfa Aesar™
A1564436
500g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

Applications
Methyltriphenylphosphonium iodide is as a reactant for synthesis of triphenylamine-based dyes and polyolefinic aromatic molecules with pyrene for use in dye-sensitized solar cells. Used as ligand in coupling reaction. It is also used as a precursor to a Wittig reagent.

Solubility
Soluble in water, acetone, dichloromethane , methanol.

Notes
Light sensitive and hygroscopic. Store in dark. Protect from humidity and water.
Description & Specifications

Chemical Identifiers

2065-66-9
404.23
UN3464
CH3P(C6H5)3I
MFCD00066175
Methyltriphenylphosphonium iodide

Specifications

2065-66-9
404.23
UN3464
25g
Methyltriphenylphosphonium iodide
CH3P(C6H5)3I
MFCD00066175
185° to 188°C
Odorless
98%

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280h-P309+P311
Wear protective gloves/clothing.
IF exposed or if you feel unwell: Call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOPHOSPHORUS COMPOUND, TOXIC, SOLID, N.O.S.