missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyltriphenylphosphonium iodide, 98%

Manufacturer:  Alfa Aesar™ A1564436

 View more versions of this product

Catalog No. AAA1564436



Description

Description

Applications
Methyltriphenylphosphonium iodide is as a reactant for synthesis of triphenylamine-based dyes and polyolefinic aromatic molecules with pyrene for use in dye-sensitized solar cells. Used as ligand in coupling reaction. It is also used as a precursor to a Wittig reagent.

Solubility
Soluble in water, acetone, dichloromethane , methanol.

Notes
Light sensitive and hygroscopic. Store in dark. Protect from humidity and water.
Specifications

Specifications

Methyltriphenylphosphonium iodide
185°C to 188°C
2065-66-9
MFCD00066175
methyltriphenylphosphonium iodide, methyl triphenylphosphonium iodide, methyltriphenylphosphanium iodide, methyl triphenyl phosphonium iodide, phosphonium, methyltriphenyl-, iodide, methyl triphenylphosphoniumiodide, methyl triphenyl phosphanium iodide, methyltriphenylphosphonium iodide, triphenylmethyl phosphonium iodide, pubchem15730
C[P+](C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3.[I-]
404.231
404.23
Odorless
500g
C19H18IP
UN3464
JNMIXMFEVJHFNY-UHFFFAOYSA-M
methyl(triphenyl)phosphanium;iodide
638159
98%

Chemical Identifiers

2065-66-9
404.231
JNMIXMFEVJHFNY-UHFFFAOYSA-M
638159
C[P+](C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3.[I-]
C19H18IP
MFCD00066175
methyltriphenylphosphonium iodide, methyl triphenylphosphonium iodide, methyltriphenylphosphanium iodide, methyl triphenyl phosphonium iodide, phosphonium, methyltriphenyl-, iodide, methyl triphenylphosphoniumiodide, methyl triphenyl phosphanium iodide, methyltriphenylphosphonium iodide, triphenylmethyl phosphonium iodide, pubchem15730
methyl(triphenyl)phosphanium;iodide
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280h-P309+P311
Wear protective gloves/clothing.
IF exposed or if you feel unwell: Call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOPHOSPHORUS COMPOUND, TOXIC, SOLID, N.O.S.

Documents