Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyltriphenylarsonium iodide, 98%

Chemical Identifiers

Chemical Name or Material Methyltriphenylarsonium iodide
CAS 1499-33-8
Molecular Formula (C6H5)3AsCH3I
Formula Weight 448.18
MDL Number MFCD00031543
UN Number UN3465
Beilstein 3639838
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1859906

 
alfa aesar™
A1859906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 448.18
Chemical Name or Material Methyltriphenylarsonium iodide
TSCA TSCA
Assay 98%
Molecular Formula (C6H5)3AsCH3I
MDL Number MFCD00031543
CAS 1499-33-8
UN Number UN3465
Beilstein 3639838
Health Hazard 1 Danger
Melting Point 173° to 177°C
Quantity 5g
Health Hazard 2 GHS H Statement
H301-H331
Toxic if swallowed.
Toxic if inhaled.
Health Hazard 3 GHS P Statement
P261-P301+P310-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOARSENIC COMPOUND, SOLID, N.O.S.
Safety and Handling

Danger

GHS P Statement
P261-P301+P310-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H331
Toxic if swallowed.
Toxic if inhaled.