Please login to your online account to display your discounted pricing

Methyltrioxorhenium (VII), 98%, ACROS Organics™


Quantity 500mg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC346965000

 View more versions of this product

Catalog No. AC346965000

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Crystals
Formula Weight 249.23
Quantity 500mg
Color White to blue-gray
Merck Index 15, 6206
CAS Max % 100.0
CAS Min % 97.5
Elemental Analysis 97.5% min.
Chemical Name or Material Methyltrioxorhenium (VII)
Infrared Spectrum Authentic
Packaging Glass bottle
Assay Percent Range 98%
Molecular Formula CH3O3Re
MDL Number MFCD00192332
CAS 70197-13-6
Synonym MTO

Specifications

Chemical Name or Material Methyltrioxorhenium (VII)
CAS 70197-13-6
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula CH3O3Re
Formula Weight 249.23
MDL Number MFCD00192332
Synonym MTO