Please login to your online account to display your discounted pricing

Methyltrioxorhenium (VII), 98%, ACROS Organics™

$133.48 - $133.48

Chemical Identifiers

Chemical Name or Material Methyltrioxorhenium (VII)
CAS 70197-13-6
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula CH3O3Re
Formula Weight 249.23
MDL Number MFCD00192332
Synonym MTO
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC346960025

  SDS
thermo fisher scientific
AC346960025
Glass bottle 2.5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC346965000

  SDS
thermo fisher scientific
AC346965000
Glass bottle 500mg Each for $133.48

AC346961000

  SDS
fisher scientific acros
346961000
Glass bottle 100mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 249.23
Physical Form Crystals
Quantity 2.5g
Color White to blue-gray
Merck Index 15, 6206
CAS Max % 100.0
Synonym MTO
Elemental Analysis 97.5% min.
Packaging Glass bottle
CAS Min % 97.5
Infrared Spectrum Authentic
CAS 70197-13-6
Chemical Name or Material Methyltrioxorhenium (VII)
MDL Number MFCD00192332
Molecular Formula CH3O3Re
Assay Percent Range 98%