missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyltrioxorhenium (VII), 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 346960025
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Methyltrioxorhenium (VII) | |
97.5 | |
2.5g | |
CH3O3Re | |
15, 6206 | |
Solubility in water: very soluble in h2o | |
[CH3-].O=[Re](=O)=O | |
249.23 | |
249.23 | |
Authentic |
70197-13-6 | |
100.0 | |
97.5% min. (GC) | |
MFCD00192332 | |
methyltrioxorhenium vii, methyltrioxorhenium, methylrhenium trioxide, unii-883d8rdd5q, methylrhenium vii trioxide, mto, methyltrioxorhenium mi, t-4-methyltrioxorhenium, methylrhenium trioxide vii | |
VZSXFJPZOCRDPW-UHFFFAOYSA-N | |
carbanide;trioxorhenium | |
2734010 | |
98% | |
Glass bottle |
RUO â Research Use Only