Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyltrioxorhenium(VII), 98%

Chemical Identifiers

Chemical Name or Material Methyltrioxorhenium(VII)
CAS 70197-13-6
Molecular Formula CH3O3Re
Linear Formula CH3ReO3
Formula Weight 249.24
MDL Number MFCD00192332
UN Number UN1479
Beilstein 3927932
Synonym MTO
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL18478MC

 
alfa aesar™
L18478MC
100mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL18478ME

 
alfa aesar™
L18478ME
500mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 249.24
Assay 98%
Molecular Formula CH3O3Re
MDL Number MFCD00192332
UN Number UN1479
Beilstein 3927932
Synonym MTO
Quantity 100mg
Health Hazard 2 GHS H Statement
H272
May intensify fire; oxidizer.
Health Hazard 3 GHS P Statement
P221-P210-P220-P280-P370+P378a-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Keep/Store away from clothing/combustible materials.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
DOT Information Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING SOLID, N.O.S.
Chemical Name or Material Methyltrioxorhenium(VII)
Linear Formula CH3ReO3
CAS 70197-13-6
Health Hazard 1 Danger
Melting Point 110° to 115°C

Applications
Methyltrioxorhenium(VII) is used as a catalyst for various transformations such as olefin metathesis. It is associated with hydrogen peroxide and used as an oxidizing agent. It is used to prepare acid or ester from the corresponding terminalalkynes. It is a catalytic oxidant used with urea hydrogen peroxide to prepare nitrones. It reacts with 2-methylnaphthalene to get 2-methyl-1,4-naphthoquinone (Vitamin K3). Further, it catalyzes the reaction with aldehydes and diazoketones to get an alkene. In addition to this, it is used as an effective antioxidant under various conditions.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P221-P210-P220-P280-P370+P378a-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Keep/Store away from clothing/combustible materials.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H272
May intensify fire; oxidizer.

2° to 8°C