missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyltrimethoxysilane, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 250540500

 View more versions of this product

Catalog No. AC250540500



Specifications

Specifications

Methyltrimethoxysilane
11°C
Glass bottle
0.95
102°C
-50°C
97%
96.0
97%
CH3Si(OCH3)3
04, 4203
Solubility in water: decomposes
CO[Si](C)(OC)OC
136.22
136.22
97%
0.9500g/mL
Authentic
1.3695 to 1.3715
0.5 mPa.s (20°C)
Colorless to Yellow
50mL
1185-55-3
100.0
C4H12O3Si
MFCD00008342
dynasylan mtms, methyl trimethoxysilane, methyl-trimethoxysilane, methyltrimethoxysilane, silane a-163, silane, methyltrimethoxy, silane, trimethoxymethyl, trimethoxy methyl silane, unii-0hi0d71mci, union carbide a-163
BFXIKLCIZHOAAZ-UHFFFAOYSA-N
trimethoxy(methyl)silane
14456
Liquid

Chemical Identifiers

1185-55-3
136.22
BFXIKLCIZHOAAZ-UHFFFAOYSA-N
14456
CO[Si](C)(OC)OC
C4H12O3Si
MFCD00008342
dynasylan mtms, methyl trimethoxysilane, methyl-trimethoxysilane, methyltrimethoxysilane, silane a-163, silane, methyltrimethoxy, silane, trimethoxymethyl, trimethoxy methyl silane, unii-0hi0d71mci, union carbide a-163
trimethoxy(methyl)silane
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Highly flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 214-685-0

RTECSNumber : VV4650000

TSCA : TSCA

SDS Documents