Please login to your online account to display your discounted pricing

Methyltributylammonium chloride, 98%, ACROS Organics™


Quantity 250g

Manufacturer: fisher scientific acros  377832500

 View more versions of this product

Catalog No. AC377832500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Infrared Spectrum Authentic
Water 2% Max.
Color Slightly pale yellow
Quantity 250g
Chemical Name or Material Methyltributylammonium chloride
CAS 56375-79-2
CAS Min % 97.5
CAS Max % 100.0
Physical Form Slightly pale yellow crystalline powder
Assay 97.5% min. (on dry substance)
Assay Percent Range 98%
Molecular Formula C13H30ClN
Formula Weight 235.84g/mol
MDL Number MFCD00011847
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Methyltributylammonium chloride
CAS 56375-79-2
CAS Min % 97.5
CAS Max % 100.0
Assay 97.5% min. (on dry substance)
Assay Percent Range 98%
Molecular Formula C13H30ClN
Formula Weight 235.84g/mol
MDL Number MFCD00011847

Phase transfer catalyst

Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

SDS & Safety Documents