Please login to your online account to display your discounted pricing

Methyltributylammonium chloride, 98%, ACROS Organics™


Quantity 1kg

Manufacturer: thermo fisher scientific  AC377830010

 View more versions of this product

Catalog No. AC377830010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5% (on dry substance)
Physical Form Crystalline Powder
Formula Weight 235.84
Quantity 1kg
Color Slightly pale yellow
Water 2% Max.
CAS Max % 100.0
Chemical Name or Material Methyltributylammonium chloride
Infrared Spectrum Authentic
Assay Percent Range 98%
MDL Number MFCD00011847
CAS 56375-79-2
CAS Min % 97.5
Molecular Formula C13H30ClN
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Methyltributylammonium chloride
CAS 56375-79-2
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5% (on dry substance)
Assay Percent Range 98%
Molecular Formula C13H30ClN
Formula Weight 235.84
MDL Number MFCD00011847
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

SDS & Safety Documents