Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyltri-n-butylammonium bromide, 98%

$57.57 - $584.62

Chemical Identifiers

Chemical Name or Material Methyltri-n-butylammonium bromide
CAS 37026-88-3
Molecular Formula [CH3(CH2)3]3NCH3Br
Formula Weight 280.29
MDL Number MFCD00043201
Beilstein 3704268
Synonym Tri-n-butylmethylammonium bromide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2382809

  SDS
alfa aesar™
B2382809
10g Each for $57.57

AAB2382818

  SDS
alfa aesar™
B2382818
50g Each for $162.73

AAB2382830

  SDS
alfa aesar™
B2382830
250g Each for $584.62
Description & Specifications

Specifications

Formula Weight 280.29
Quantity 10g
Synonym Tri-n-butylmethylammonium bromide
Beilstein 3704268
Assay 98%
Chemical Name or Material Methyltri-n-butylammonium bromide
Melting Point 120° to 122°C
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula [CH3(CH2)3]3NCH3Br
MDL Number MFCD00043201
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Warning
CAS 37026-88-3
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.