missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyltin trichloride, 97%

Manufacturer:  Alfa Aesar™ 7115614

 View more versions of this product

Catalog No. AA7115614



Specifications

Specifications

Methyltin trichloride
993-16-8
Moisture Sensitive
MFCD00000514
methyltin trichloride, methyltrichlorotin, stannane, trichloromethyl, trichloromethyltin, monomethyltin trichloride, trichloro methyl stannane, methyltrichlorostannane, tin, methyl-, trichloride, unii-q2e6v14u06, ccris 6327
C[Sn](Cl)(Cl)Cl
240.095
240.11
97%
43°C
25g
CH3Cl3Sn
UN2923
YFRLQYJXUZRYDN-UHFFFAOYSA-K
trichloro(methyl)stannane
101940
Crystalline

Chemical Identifiers

993-16-8
240.095
YFRLQYJXUZRYDN-UHFFFAOYSA-K
101940
C[Sn](Cl)(Cl)Cl
CH3Cl3Sn
MFCD00000514
methyltin trichloride, methyltrichlorotin, stannane, trichloromethyl, trichloromethyltin, monomethyltin trichloride, trichloro methyl stannane, methyltrichlorostannane, tin, methyl-, trichloride, unii-q2e6v14u06, ccris 6327
trichloro(methyl)stannane
Safety and Handling

Safety and Handling

GHS H Statement
H228-H300-H310-H330-H314-H318
Flammable solid.
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P210-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.

TSCA : TSCA

SDS Documents