Please login to your online account to display your discounted pricing

Methylthymol Blue, Tetrasodium Salt, Watersoluble, ACROS Organics™

$116.20 - $116.20

Chemical Identifiers

Chemical Name or Material Methylthymol Blue, tetrasodium salt
CAS 1945-77-3
Purity Grade Pure
Molecular Formula C37H40N2Na4O13S
Formula Weight 844.76
MDL Number MFCD00148905
Synonym 3,3'-Bis[N,N-di(carboxymethyl)aminomethyl]thymolsulfonephthalein, sodium salt
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC106370050

  SDS
fisher scientific acros
106370050
Glass bottle 5g Each for $116.20
Description & Specifications

Specifications

Physical Form Powder
Formula Weight 844.76
Color Dark green to reddish-brown
Quantity 5g
Merck Index 15, 6202
EINECS Number 217-743-3
TSCA TSCA
Purity Grade Pure
Beilstein 19, V,8, 619
Infrared Spectrum Authentic
Lambda Max 435 to 440nm (in water)
Loss on Drying 10% max. (110°C, 1 hr)
Specific Absorbance 130 min. min. (at lambda max, in water)
Chemical Name or Material Methylthymol Blue, tetrasodium salt
Molecular Formula C37H40N2Na4O13S
MDL Number MFCD00148905
CAS 1945-77-3
Synonym 3,3'-Bis[N,N-di(carboxymethyl)aminomethyl]thymolsulfonephthalein, sodium salt
Packaging Glass bottle

Indicator