Please login to your online account to display your discounted pricing

Alfa Aesar™ Methylthiomethyl p-tolyl sulfone, 99%

$109.74 - $364.81

Chemical Identifiers

Chemical Name or Material Methylthiomethyl p-tolyl sulfone
CAS 59662-65-6
Molecular Formula C9H12O2S2
Formula Weight 216.32
MDL Number MFCD00010750
Synonym 4-[(Methylthio)methylsulfonyl]toluene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3187706

 
alfa aesar™
H3187706
5g Each for $109.74

AAH3187714

 
alfa aesar™
H3187714
25g Each for $364.81
Description & Specifications

Specifications

Formula Weight 216.32
Chemical Name or Material Methylthiomethyl p-tolyl sulfone
Assay 99%
Molecular Formula C9H12O2S2
MDL Number MFCD00010750
CAS 59662-65-6
Synonym 4-[(Methylthio)methylsulfonyl]toluene
Melting Point 77° to 81°C
Quantity 5g