Please login to your online account to display your discounted pricing

Alfa Aesar™ Methylthiomethyl acetate, 95%

Chemical Identifiers

Chemical Name or Material Methylthiomethyl acetate
CAS 16437-69-7
Molecular Formula C4H8O2S
Linear Formula CH3CO2CH2SCH3
Formula Weight 120.17
MDL Number MFCD00075528
UN Number UN3272
Synonym Acetic acid methylthiomethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3231009

  SDS
alfa aesar™
H3231009
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3231018

  SDS
alfa aesar™
H3231018
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 120.17
Health Hazard 2 GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 10g
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
Health Hazard 1 Warning
Linear Formula CH3CO2CH2SCH3
Health Hazard 3 GHS P Statement
P261-P280g-P305+P351+P338
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Chemical Name or Material Methylthiomethyl acetate
MDL Number MFCD00075528
Boiling Point 149°C
CAS 16437-69-7
Flash Point 44°C (111°F)
Molecular Formula C4H8O2S
Synonym Acetic acid methylthiomethyl ester
UN Number UN3272
Assay 95%
Odor Pungent
Safety and Handling

Warning

GHS P Statement
P261-P280g-P305+P351+P338
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.