Please login to your online account to display your discounted pricing

Alfa Aesar™ (Methylthio)acetonitrile, 99%

$32.01 - $106.66

Chemical Identifiers

Chemical Name or Material (Methylthio)acetonitrile
CAS 35120-10-6
Molecular Formula C3H5NS
Linear Formula CH3SCH2CN
Formula Weight 87.14
MDL Number MFCD00001925
Beilstein 1738015
Synonym (Methylmercapto)acetonitrile
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0306614

 
alfa aesar™
L0306614
25g Each for $106.66

AAL0306606

 
alfa aesar™
L0306606
5g Each for $32.01
Description & Specifications

Specifications

Formula Weight 87.14
Chemical Name or Material (Methylthio)acetonitrile
Molecular Formula C3H5NS
MDL Number MFCD00001925
CAS 35120-10-6
Assay 99%
Linear Formula CH3SCH2CN
Beilstein 1738015
Synonym (Methylmercapto)acetonitrile
Health Hazard 1 Warning
Boiling Point 166° to 167°C
Quantity 25g
Flash Point 67°C (152°F)
Health Hazard 2 GHS H Statement
H302-H312-H332-H315-H319-H335-H227
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Health Hazard 3 GHS P Statement
P260-P280h-P305+P351+P338
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Safety and Handling

Warning

GHS P Statement
P260-P280h-P305+P351+P338
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H302-H312-H332-H315-H319-H335-H227
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.